Learn More
AdipoGen (-)-Ageloxime D (1 mg)
SDP
Catalog No. 502053362
Encompass_Preferred
Chemical. CAS: 1219817-25-0. Formula: C26H40N5O. MW: 438.6. Isolated from sponge Agelas sp. Quaternary ammonium compound. Antifouling compound. Inhibits biofilm formation but not bacterial growth of Staphylococcus epidermidis. Anticancer compound. Cytotoxic against L5178Y mouse lymphoma cells.
Catalog No. 50-205-3362 Supplier AdipoGen Supplier No. AGCN20016M001
Chemical. CAS: 1219817-25-0. Formula: C26H40N5O. MW: 438.6. Isolated from sponge Agelas sp. Quaternary ammonium compound. Antifouling compound. Inhibits biofilm formation but not bacterial growth of Staphylococcus epidermidis. Anticancer compound. Cytotoxic against L5178Y mouse lymphoma cells.