Learn More
Beckman Coulter DNAdvance Kit, 384 Preps
Catalog No. NC1214906
Encompass
Isolation from mammalian tissue. Built on SPRI paramagnetic bead-based technology, the DNAdvance Kit is a high throughput genomic DNA (gDNA) isolation reagent kit enabling the purification of high quality DNA from mammalian tissue samples. The DNAdvance Kit offers greater recovery of high quality gDNA compared to equivalent DNA isolation kits from competitors.
This item is not returnable. View return policy
Catalog No. NC1214906 Supplier Beckman Coulter Supplier No. A48705
Isolation from mammalian tissue. Built on SPRI paramagnetic bead-based technology, the DNAdvance Kit is a high throughput genomic DNA (gDNA) isolation reagent kit enabling the purification of high quality DNA from mammalian tissue samples. The DNAdvance Kit offers greater recovery of high quality gDNA compared to equivalent DNA isolation kits from competitors.