Learn More
Aviva Systems Biology Corp AGER Antibody - C-terminal region (ARP41643_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41643_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AGER. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human AGER. Alternate names: MGC22357, RAGEApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57932