Learn More
Medchemexpress LLC HY-P7467 10ug Medchemexpress, AGER, His (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP746710UG
Encompass_Preferred
Medchemexpress, HY-P7467 10ug AGER, His (HEK293-expressed), Human CAS: Recombinant Human AGER, His (HEK293-expressed) is human recombinant AGER with a His tag at the C-terminus. Recombinant Human AGER, His (HEK293-expressed) is expressed by mammalian expression system and the target gene encoding Ala23-Ala344 is expressed. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3168