Learn More
ABclonal Technology AGER Rabbit mAb
SDP
Catalog No. 5000005374
Encompass_Preferred
The advanced glycosylation end product (AGE) receptor encoded by this gene is a member of the immunoglobulin superfamily of cell surface receptors It is a multiligand receptor and besides AGE interacts with other molecules implicated in homeostasis development and inflammation and certain diseases such as diabetes and Alzheimer s disease Many alternatively spliced transcript variants encoding different isoforms as well as non-protein-coding variants have been described for this gene (PMID 18089847)
Catalog No. 50-000-05374 Supplier ABclonal Technology Supplier No. A23422100UL
The advanced glycosylation end product (AGE) receptor encoded by this gene is a member of the immunoglobulin superfamily of cell surface receptors It is a multiligand receptor and besides AGE interacts with other molecules implicated in homeostasis development and inflammation and certain diseases such as diabetes and Alzheimer s disease Many alternatively spliced transcript variants encoding different isoforms as well as non-protein-coding variants have been described for this gene (PMID 18089847)