Learn More
Medchemexpress LLC HY-15200 10mg Medchemexpress, Agerafenib CAS:1188910-76-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1520010MG
Encompass_Preferred
Medchemexpress, HY-15200 10mg Agerafenib CAS:1188910-76-0 Agerafenib (CEP-32496; RXDX-105) is a highly potent and orally efficacious inhibitor of BRAFV600E with a Kd of 14 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9095