Learn More
Adam Equipment AGF 1320a Floor Scale 1320lb / 600kg capacity x 0.05lb / 0.02kg readability
Catalog No. NC2611856
Encompass_Preferred
Adam Equipment AGF floor Scales are Ideal for industrial weighing and counting, AGB and AGF bench and floor scales feature sturdy construction and a rugged stainless steel indicator. The scales are available in a choice of platform sizes and a wide selection of capacities. Operation is fast and easy, as results are readily visible on the backlit LCD that boasts 1.5in / 40mm-high digits. An internal rechargeable battery means the AGB/AGF can be used in locations where power is unavailable. Swiftly transmit results to computers or printers using the RS-232 interface. Versatile and easy to operate, AGB and AGF scales are well-suited for use in warehouses, shipping and receiving areas, factories and production facilities. The scales offer capacities from 8kg to 600kg and perform a variety of weighing and counting tasks. Featuring an internal rechargeable battery, the AGB/AGF can be used in locations where power is unavailable. Rechargeable battery included for operation almost anywhere.
Catalog No. NC2611856 Supplier Adam Equipment Supplier No. AGF 1320A
Adam Equipment AGF floor Scales are Ideal for industrial weighing and counting, AGB and AGF bench and floor scales feature sturdy construction and a rugged stainless steel indicator. The scales are available in a choice of platform sizes and a wide selection of capacities. Operation is fast and easy, as results are readily visible on the backlit LCD that boasts 1.5in / 40mm-high digits. An internal rechargeable battery means the AGB/AGF can be used in locations where power is unavailable. Swiftly transmit results to computers or printers using the RS-232 interface. Versatile and easy to operate, AGB and AGF scales are well-suited for use in warehouses, shipping and receiving areas, factories and production facilities. The scales offer capacities from 8kg to 600kg and perform a variety of weighing and counting tasks. Featuring an internal rechargeable battery, the AGB/AGF can be used in locations where power is unavailable. Rechargeable battery included for operation almost anywhere.