Learn More
Aviva Systems Biology Corp AGGF1 Antibody - middle region (ARP57098_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57098_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AGGF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AGGF1. Alternate names: FLJ10283, GPATC7, GPATCH7, HSU84971, HUS84971, VG5QApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62862