Learn More
Biomatik Corporation Aggrecan Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50503
Encompass_Preferred
Aggrecan Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Aggrecan. The Aggrecan Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 1~300aa. Applications: ELISA, WB, IHC. Target Synonyms: Aggrecan. Accession Number: P16112. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6443