Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Aggrecan (PG) ELISA Kit

Catalog No. KAP1461
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KAP1461 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KAP1461 Supplier Invitrogen™ Supplier No. KAP1461
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Aggrecan (PG) ELISA quantitates Hu Aggrecan (PG) in humansynovial fluid, serum, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Aggrecan (PG). Principle of the method The Human Aggrecan (PG) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second enzyme-conjugated (detector) antibody, binding to the target on a different epitope from the capture antibody. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluronan binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16112
Assay Range 10-250 ng/mL
Assay Sensitivity 0.9 ng/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody(s), diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Serum, Supernatant, Synovial Fluid
Substrate Type Proteoglycans
Target Species Human
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ACAN; Agc; Agc1; AGCAN; aggrecan; aggrecan 1; aggrecan 1 (chondroitin sulfate proteoglycan 1, large aggregating proteoglycan, antigen identified by monoclonal antibody A0122); aggrecan core protein; Aggrecan core protein 2; aggrecan CS2 domain; aggrecan epidermal growth factor-like domain-1; aggrecan G2 domain; aggrecan interglobular domain; aggrecan keratan sulfate domain; aggrecan, structural proteoglycan of cartilage; articular cartilage aggrecan precursor; b2b183Clo; cartilage aggregating proteoglycan; cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; chondroitin sulfate proteoglycan core protein 1; cmd; CSPCP; CSPG1; CSPGCP; high density cartilage proteoglycan; large aggregating cartilage proteoglycan core protein; large aggregating proteoglycan; large aggregating proteoglycan, antigen; MSK16; SEDKACAN; Agc; Agc1; AGCAN; aggrecan; aggrecan 1; aggrecan 1 (chondroitin sulfate proteoglycan 1, large aggregating proteoglycan, antigen identified by monoclonal antibody A0122); aggrecan core protein; Aggrecan core protein 2; aggrecan CS2 domain; aggrecan epidermal growth factor-like domain-1; aggrecan G2 domain; aggrecan interglobular domain; aggrecan keratan sulfate domain; aggrecan, structural proteoglycan of cartilage; articular cartilage aggrecan precursor; b2b183Clo; cartilage aggregating proteoglycan; cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; chondroitin sulfate proteoglycan core protein 1; cmd; CSPCP; CSPG1; CSPGCP; high density cartilage proteoglycan; large aggregating cartilage proteoglycan core protein; large aggregating proteoglycan; large aggregating proteoglycan, antigen; MSK16; SEDK; AGC1; AGCAN; CSPG1; CSPGCP; MSK16; SEDK
Gene ID (Entrez) 176
Gene Symbol ACAN
Incubation Time 3.25 hr.
Interassay CV 5.6%
Intraassay CV 2.1%
Kit Contents Pre-coated 96 well plate, Standard, Sample diluent, Incubation Buffer, HRP conjugated Detection Antibody, Conjugate Diluent, Wash Buffer, Chromogen, Stop Solution, Controls, Standard, Sample diluent, Incubation Buffer, HRP conjugated Detection Antibody, Conjugate Diluent, Wash Buffer, Chromogen, Stop Solution, Controls
Label or Dye HRP
Marker Gene PG
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Serum 20 μL, Supernatant 10 μL, Synovial Fluid 25 μL
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target Aggrecan
Test Time 1 hr 20 min
Total Assay Time 3 hr 15 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.