Learn More
AnaSpec AGGRESUREB-AMYLOID (1-42), HUM
Supplier:  AnaSpec AS72216
Encompass
AggreSure(TM) B -Amyloid (1-42), Human[DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA] ; MW 4514.1; (0.25 mg)
Catalog No. NC0721474