Learn More
STEMCELL Technologies AggreWell™ EB Formation Medium, Size: 100 mL
SDP
Catalog No. NC0488033
Encompass
AggreWell™ EB Formation Medium is a serum-free medium that supports the survival of mTeSR™1-, TeSR™2-, or TeSR™-E8™-cultured human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells during generation and subsequent culture of embryoid bodies (EBs).
Catalog No. NC0488033 Supplier STEMCELL Technologies Supplier No. 05893
AggreWell™ EB Formation Medium is a serum-free medium that supports the survival of mTeSR™1-, TeSR™2-, or TeSR™-E8™-cultured human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells during generation and subsequent culture of embryoid bodies (EBs).