Learn More
Selleck Chemical LLC AGI 1067
SDP
Catalog No. 5000004691
Encompass_Preferred
AGI-1067 is a novel phenolic intra- and extracellular antioxidant that inhibits the expression of a number of proinflammatory genes involved in atherosclerosis
Catalog No. 50-000-04691 Supplier Selleck Chemical LLC Supplier No. S64205MG
AGI-1067 is a novel phenolic intra- and extracellular antioxidant that inhibits the expression of a number of proinflammatory genes involved in atherosclerosis