Learn More
Medchemexpress LLC HY-112130 10mg Medchemexpress, AGI-24512 CAS:2201066-53-5 Purity:>98%
SDP
Catalog No. 501969030
Encompass_Preferred
Medchemexpress, HY-112130 10mg AGI-24512 CAS:2201066-53-5 AGI-24512 is a methionine adenosyltransferase 2A (MATA2 ) inhibitors useful for treatment of cancer. AGI-24512 blocks growth of MTAP-deleted cancer cells in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9030 Supplier Medchemexpress LLC Supplier No. HY11213010MG
Medchemexpress, HY-112130 10mg AGI-24512 CAS:2201066-53-5 AGI-24512 is a methionine adenosyltransferase 2A (MATA2 ) inhibitors useful for treatment of cancer. AGI-24512 blocks growth of MTAP-deleted cancer cells in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.