Learn More
Medchemexpress LLC Agi-24512 | 2201066-53-5 | 99.5% | 400.47 | 1 ML
SDP
Catalog No. 5000437957
Encompass_Preferred
AGI-24512 is a potent methionine adenosyltransferase 2α (MAT2A) inhibitor with an IC50 of 8 nM. It triggers DNA damage response and can block the proliferation of MTAP-deleted cancer cells in vitro. AGI-24512 can be used for researching anticancer properties.
  • Triggers DNA damage response
  • Blocks proliferation of MTAP-deleted cancer cells in vitro
  • Inhibits PRMT5-mediated SDMA marks
  • Leads to a dose-dependent decrease in SAM levels
  • Suitable for anticancer research
Catalog No. 50-004-37957 Supplier Medchemexpress LLC Supplier No. HY11213010MM/1ML
AGI-24512 is a potent methionine adenosyltransferase 2α (MAT2A) inhibitor with an IC50 of 8 nM. It triggers DNA damage response and can block the proliferation of MTAP-deleted cancer cells in vitro. AGI-24512 can be used for researching anticancer properties.
  • Triggers DNA damage response
  • Blocks proliferation of MTAP-deleted cancer cells in vitro
  • Inhibits PRMT5-mediated SDMA marks
  • Leads to a dose-dependent decrease in SAM levels
  • Suitable for anticancer research