Learn More
TARGETMOL CHEMICALS INC AGI-43192 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T97295MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AGI-43192 is a potent methionine adenosine transferase 2A (MAT2A) inhibitor that penetrates the blood-brain barrier. AGI-43192 inhibits S-adenosine methionine (SAM) production metabolic enzyme and methionine adenosine transferase 2A (MAT2A) which has the potential to study the role of SAM regulation in the central nervous system (CNS) and the treatment of cancer diseases. purity: 99%
Catalog No. 50-257-8968