Learn More
TARGETMOL CHEMICALS INC AGI-5198 100MG
SDP
Catalog No. 502577225
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AGI-5198 (IDH-C35) is a highly effective and specific inhibitor of IDH1 R132H/R132C mutants (IC50 0.07/0.16 uM). purity: 99%
Catalog No. 50-257-7225 Supplier TARGETMOL CHEMICALS INC Supplier No. T2104100MG
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AGI-5198 (IDH-C35) is a highly effective and specific inhibitor of IDH1 R132H/R132C mutants (IC50 0.07/0.16 uM). purity: 99%