Learn More
Medchemexpress LLC AGI-5198 | 1355326-35-0 | 99.9% | 462.56 | 5 MG
SDP
Catalog No. 5000386949
Encompass_Preferred
AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM. It causes 50 to 60% growth inhibition of human glioma xenografts and does not affect the growth of IDH1 wild-type glioma xenografts. This compound reverses the effects of ionizing radiation (IR) on IDH1(R132H) mutated cancer cells, leading to higher levels of reactive oxygen species, DNA double-strand breaks, and cell death.
  • Potent and selective mutant IDH1R132H inhibitor.
  • Causes 50 to 60% growth inhibition of human glioma xenografts.
  • Does not affect the growth of IDH1 wild-type glioma xenografts.
  • Reverses the effects of ionizing radiation (IR) on IDH1(R132H) mutated cancer cells, leading to higher levels of reactive oxygen species, DNA double-strand breaks, and cell death.
Catalog No. 50-003-86949 Supplier Medchemexpress LLC Supplier No. HY180825MG
Add to Cart
Add to Cart
AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM. It causes 50 to 60% growth inhibition of human glioma xenografts and does not affect the growth of IDH1 wild-type glioma xenografts. This compound reverses the effects of ionizing radiation (IR) on IDH1(R132H) mutated cancer cells, leading to higher levels of reactive oxygen species, DNA double-strand breaks, and cell death.
  • Potent and selective mutant IDH1R132H inhibitor.
  • Causes 50 to 60% growth inhibition of human glioma xenografts.
  • Does not affect the growth of IDH1 wild-type glioma xenografts.
  • Reverses the effects of ionizing radiation (IR) on IDH1(R132H) mutated cancer cells, leading to higher levels of reactive oxygen species, DNA double-strand breaks, and cell death.