Learn More
Selleck Chemical LLC AGI-5198
SDP
Catalog No. 5000004331
Encompass_Preferred
AGI-5198 (IDH-C35) is the first highly potent and selective inhibitor of IDH1 R132H/R132C mutants with IC50 of 0 07 M/0 16 M
Catalog No. 50-000-04331 Supplier Selleck Chemical LLC Supplier No. S71855MG
AGI-5198 (IDH-C35) is the first highly potent and selective inhibitor of IDH1 R132H/R132C mutants with IC50 of 0 07 M/0 16 M