Learn More
Medchemexpress LLC HY-18082 100mg , AGI-5198 IDH-C35 CAS:1355326-35-0 Purity:98%
SDP
Catalog No. 501492727
Encompass_Preferred
Medchemexpress, HY-18082 100mg AGI-5198 IDH-C35 CAS:1355326-35-0 Formula:C27H31FN4O2 Purity:98% AGI-5198 is a novel R132H-IDH1 inhibitor, used for cancer treatment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2727 Supplier Medchemexpress LLC Supplier No. HY18082100MG
Medchemexpress, HY-18082 100mg AGI-5198 IDH-C35 CAS:1355326-35-0 Formula:C27H31FN4O2 Purity:98% AGI-5198 is a novel R132H-IDH1 inhibitor, used for cancer treatment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.