Learn More
Medchemexpress LLC AGI-5198 (IDH-C35) | 1355326-35-0 | 99.9% | 462.56 | 1 G
SDP
Catalog No. 5000387205
Encompass_Preferred
AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor, with an IC50 of 0.07 μM. It is for research use only and demonstrates dose-dependent inhibition of the mutant IDH1 enzyme in TS603 glioma cells. AGI-5198 can also reverse the effects of increased reactive oxygen species, DNA double-strand breaks, and cell death in IDH1(R132H) mutated cancer cells after exposure to ionizing radiation. In vivo studies show it causes 50 to 60% growth inhibition of human glioma xenografts.
  • Solid appearance with a white to off-white color
  • Inhibits IDH1R132H mutant with an IC50 of 0.07 μM
  • Soluble in DMF and DMSO for in vitro applications
  • Available in powder and solvent forms for varied storage conditions
  • Demonstrates significant growth inhibition in human glioma xenografts
Catalog No. 50-003-87205 Supplier Medchemexpress LLC Supplier No. HY180821G
AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor, with an IC50 of 0.07 μM. It is for research use only and demonstrates dose-dependent inhibition of the mutant IDH1 enzyme in TS603 glioma cells. AGI-5198 can also reverse the effects of increased reactive oxygen species, DNA double-strand breaks, and cell death in IDH1(R132H) mutated cancer cells after exposure to ionizing radiation. In vivo studies show it causes 50 to 60% growth inhibition of human glioma xenografts.
  • Solid appearance with a white to off-white color
  • Inhibits IDH1R132H mutant with an IC50 of 0.07 μM
  • Soluble in DMF and DMSO for in vitro applications
  • Available in powder and solvent forms for varied storage conditions
  • Demonstrates significant growth inhibition in human glioma xenografts