Learn More
Medchemexpress LLC Agi-5198 (idh-c35) | 1355326-35-0 | 99.9% | 462.56 | 1 ML
SDP
Catalog No. 5000387204
Encompass_Preferred
AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM.
  • Demonstrates dose-dependent inhibition of mutant IDH1 enzyme in TS603 glioma cells.
  • Does not impair colony formation of patient-derived glioma lines expressing only wild-type IDH1.
  • Reverses increased reactive oxygen species, DNA double-strand breaks, and cell death in IDH1(R132H) cancer cells exposed to ionizing radiation.
  • Causes 50 to 60% growth inhibition of human glioma xenografts in vivo.
  • Reduces Ki-67 protein staining in tumors from treated mice.
  • Does not affect the growth of IDH1 wild-type glioma xenografts.
Catalog No. 50-003-87204 Supplier Medchemexpress LLC Supplier No. HY1808210MM/1ML
AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM.
  • Demonstrates dose-dependent inhibition of mutant IDH1 enzyme in TS603 glioma cells.
  • Does not impair colony formation of patient-derived glioma lines expressing only wild-type IDH1.
  • Reverses increased reactive oxygen species, DNA double-strand breaks, and cell death in IDH1(R132H) cancer cells exposed to ionizing radiation.
  • Causes 50 to 60% growth inhibition of human glioma xenografts in vivo.
  • Reduces Ki-67 protein staining in tumors from treated mice.
  • Does not affect the growth of IDH1 wild-type glioma xenografts.