Learn More
TARGETMOL CHEMICALS INC AGI-6780 10MG
SDP
Catalog No. 502577295
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AGI-6780 is a potent and selective inhibitor of IDH2 R140Q mutant. purity: 98%
Catalog No. 50-257-7295 Supplier TARGETMOL CHEMICALS INC Supplier No. T180910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AGI-6780 is a potent and selective inhibitor of IDH2 R140Q mutant. purity: 98%