Learn More
Apexbio Technology LLC AGI-6780 1432660-47-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000095800
Encompass_Preferred
AGI-6780 (CAS 1432660-47-3) is a potent and selective inhibitor of the oncogenic IDH2/R140Q mutant enzyme exhibiting an IC50 of 23 nM In cellular assays AGI-6780 effectively reduces intracellular 2-hydroxyglutarate (2HG) levels in TF1/R140Q human glioblastoma cells The compound also reverses the differentiation block mediated by IDH2/R140Q in TF1 cells in the presence of erythropoietin restoring erythroid marker expression such as KLF1 and Hemoglobin G1/2 AGI-6780 serves as a valuable tool for studying mutant IDH2-driven metabolic alterations and cellular differentiation processes in cancer research
Catalog No. 50-000-95800 Supplier Apexbio Technology LLC Supplier No. A31465.1
AGI-6780 (CAS 1432660-47-3) is a potent and selective inhibitor of the oncogenic IDH2/R140Q mutant enzyme exhibiting an IC50 of 23 nM In cellular assays AGI-6780 effectively reduces intracellular 2-hydroxyglutarate (2HG) levels in TF1/R140Q human glioblastoma cells The compound also reverses the differentiation block mediated by IDH2/R140Q in TF1 cells in the presence of erythropoietin restoring erythroid marker expression such as KLF1 and Hemoglobin G1/2 AGI-6780 serves as a valuable tool for studying mutant IDH2-driven metabolic alterations and cellular differentiation processes in cancer research