Learn More
Medchemexpress LLC HY-15734 5mg Medchemexpress, AGI-6780 CAS:1432660-47-3 Purity:>98%
SDP
Catalog No. 502081782
Encompass_Preferred
Medchemexpress, HY-15734 5mg AGI-6780 CAS:1432660-47-3 AGI-6780 that potently and selectively inhibits the tumor-associated mutant IDH2R140Q with IC50 of 23±1.7 nM. AGI-6780 is less potent against IDH2WT with IC50 of 190±8.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1782 Supplier Medchemexpress LLC Supplier No. HY157345MG
Medchemexpress, HY-15734 5mg AGI-6780 CAS:1432660-47-3 AGI-6780 that potently and selectively inhibits the tumor-associated mutant IDH2R140Q with IC50 of 23±1.7 nM. AGI-6780 is less potent against IDH2WT with IC50 of 190±8.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.