Learn More
Selleck Chemical LLC AGI-6780
SDP
Catalog No. 5000004042
Encompass_Preferred
AGI-6780 is a potent and selective inhibitor of IDH2 R140Q mutant with IC50 of 23 nM
Catalog No. 50-000-04042 Supplier Selleck Chemical LLC Supplier No. S724110MM/1ML
AGI-6780 is a potent and selective inhibitor of IDH2 R140Q mutant with IC50 of 23 nM