Learn More
Selleck Chemical LLC AGI-6780
SDP
Supplier:  Selleck Chemical LLC S724110MG
Encompass_Preferred
AGI-6780 is a potent and selective inhibitor of IDH2 R140Q mutant with IC50 of 23 nM
Catalog No. 50-000-04724