Learn More
Analytical Sales & Services Agilent 1050 1100 1200 1260 (1409)
Catalog No. 5000806360
Encompass
Agilent 1050 1100 1200 1260 1290 Inlet Mechanical Check Valve Assembly
Qty/Pkg: EACH
Catalog No. 50-008-06360 Supplier Analytical Sales & Services Supplier No. 92501409
Agilent 1050 1100 1200 1260 1290 Inlet Mechanical Check Valve Assembly
Qty/Pkg: EACH