Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Krackeler Scientific Adjustable and Open-ended Wrenches
SDP

Catalog No. 87101712N
Change view
Click to view available options
Length (Metric):
30.48 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
87101712N 30.48 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 87101712N Supplier Krackeler Scientific Supplier No. 9287101712EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wrenches for use with zero dead-volume fittings

  • Available in open-ended or adjustable styles
  • Available in two sizes

Specifications

Length (English) 12 in.
Length (Metric) 30.48 cm
Size 12 in.
Style Adjustable
Product Type Adjustable Wrench
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.