Learn More
Agilent Technologies AGILENT BIONYLZR TEST CHIP KIT
Catalog No. NC2388196
Encompass
Test Chip Kit. For electrophoresis instrument troubleshooting and diagnostics. Includes autofocus chip, electrode/diode test chip, and documentation.
Catalog No. NC2388196 Supplier Agilent Technologies Supplier No. G293868300
Test Chip Kit. For electrophoresis instrument troubleshooting and diagnostics. Includes autofocus chip, electrode/diode test chip, and documentation.