Learn More
Fluotics Agilent Bravo Vprep compatible P70, non-sterile, Wide Bore
Catalog No. 5000435174
Encompass_Preferred
Agilent Bravo Vprep compatible P70, non-sterile, Wide Bore, Working Volume: 70uL, Tip Color: Clear, Tip Box, 384ST/96ST, Compare To: 19134-002, 50 racks/case with 384 tips per rack
Catalog No. 50-004-35174 Supplier Fluotics Supplier No. AGI70WB.NS
Agilent Bravo Vprep compatible P70, non-sterile, Wide Bore, Working Volume: 70uL, Tip Color: Clear, Tip Box, 384ST/96ST, Compare To: 19134-002, 50 racks/case with 384 tips per rack