Learn More
Agilent Technologies Agilent Cap, Screw, Green, PTFE/Red Silicone, 9-425 Thread
Catalog No. NC3981663
Encompass
These green screw caps come with a press-fit septum made from white PTFE/red silicone bi-layer. This combination is suitable for a broad range of applications, especially HPLC applications where the sample is less volatile or where snap or crimp-style vials are not appropriate. This version is compatible with all wide-opening 2 mL glass and polypropylene vials, 2 mL micro vials, and 2 mL high recovery vials.
  • Press-fit (non-bonded) design allows for easier disposal and recycling.
  • Compatible with all wide-opening 2 mL glass and polypropylene vials, 2 mL micro vials, and 2 mL high recovery vials.
  • Cap color is green.
  • Cap material is polypropylene.
  • Cap size is 12 mm.
  • Septa layering is bi-layer.
  • Septa material is PTFE/red silicone.
  • Technique compatibility includes GC & LC.
  • Thread size is 9-425.
  • Vial type is screw.
Catalog No. NC3981663 Supplier Agilent Technologies Supplier No. 51820718
These green screw caps come with a press-fit septum made from white PTFE/red silicone bi-layer. This combination is suitable for a broad range of applications, especially HPLC applications where the sample is less volatile or where snap or crimp-style vials are not appropriate. This version is compatible with all wide-opening 2 mL glass and polypropylene vials, 2 mL micro vials, and 2 mL high recovery vials.
  • Press-fit (non-bonded) design allows for easier disposal and recycling.
  • Compatible with all wide-opening 2 mL glass and polypropylene vials, 2 mL micro vials, and 2 mL high recovery vials.
  • Cap color is green.
  • Cap material is polypropylene.
  • Cap size is 12 mm.
  • Septa layering is bi-layer.
  • Septa material is PTFE/red silicone.
  • Technique compatibility includes GC & LC.
  • Thread size is 9-425.
  • Vial type is screw.