Learn More
Agilent Technologies Agilent Caps/Septa, Crimp, 20 mm, Silver Aluminum, Bi-Layer PTFE/Silicone
Catalog No. NC3981648
Encompass
These caps/septa are designed for crimp-top vials, featuring a 20 mm silver aluminum cap and a bi-layer PTFE/silicone septa. They are suitable for applications requiring a maximum temperature of 180°C.
  • Crimp-style closure.
  • 20 mm cap size.
  • Silver aluminum cap material.
  • Bi-layer PTFE/silicone septa included.
  • Compatible with applications up to 180°C.
  • Supplied in a case of 1,000.
Catalog No. NC3981648 Supplier Agilent Technologies Supplier No. 80100191
These caps/septa are designed for crimp-top vials, featuring a 20 mm silver aluminum cap and a bi-layer PTFE/silicone septa. They are suitable for applications requiring a maximum temperature of 180°C.
  • Crimp-style closure.
  • 20 mm cap size.
  • Silver aluminum cap material.
  • Bi-layer PTFE/silicone septa included.
  • Compatible with applications up to 180°C.
  • Supplied in a case of 1,000.