Learn More
Glass Expansion Agilent 5100 / 5110 RF Coil, Silver
SDP
Catalog No. 501907335
Encompass_Preferred
RF Coil Silver for Agilent 5000 Series ICP-OES. Compatible with all Agilent 5000 Series ICP-OES instruments. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch, in addition to the power amplifier or solid-state generator. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance and gold the best corrosion resistance. For RF Coil Gold NC2081830.To install your RF coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit for Agilent 5000 Series (70-803-1415).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7335 Supplier Glass Expansion Supplier No. 709005100S
RF Coil Silver for Agilent 5000 Series ICP-OES. Compatible with all Agilent 5000 Series ICP-OES instruments. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch, in addition to the power amplifier or solid-state generator. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance and gold the best corrosion resistance. For RF Coil Gold NC2081830.To install your RF coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit for Agilent 5000 Series (70-803-1415).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001