Learn More
Agilent Technologies Agilent Crimp Cap, 11 mm, PTFE/silicone/PTFE, Septa
Catalog No. NC3981652
Encompass
This crimp cap is designed for use with 2 mL crimp top vials. It features an 11 mm aluminum cap and a tri-layer septa made of PTFE/silicone/PTFE. Each pack contains 5,000 caps.
  • Includes septa
  • Tri-layer septa layering
  • Silver cap color
  • Aluminum cap material
  • 11 mm cap size
  • Septa material: PTFE/silicone/PTFE
  • Not pre-slit septa
  • Quantity: 5,000 per pack
  • Suitable for GC, GC/MS, LC, LC/MS, and sample preparation techniques
  • Designed for crimp vial type
Catalog No. NC3981652 Supplier Agilent Technologies Supplier No. 51904051
This crimp cap is designed for use with 2 mL crimp top vials. It features an 11 mm aluminum cap and a tri-layer septa made of PTFE/silicone/PTFE. Each pack contains 5,000 caps.
  • Includes septa
  • Tri-layer septa layering
  • Silver cap color
  • Aluminum cap material
  • 11 mm cap size
  • Septa material: PTFE/silicone/PTFE
  • Not pre-slit septa
  • Quantity: 5,000 per pack
  • Suitable for GC, GC/MS, LC, LC/MS, and sample preparation techniques
  • Designed for crimp vial type