Learn More
Agilent Technologies Agilent Crimp Cap, Blue, PTFE/Silicone/PTFE, 11 mm
Catalog No. NC3981653
Encompass
This crimp cap is designed for GC and LC applications. It features a blue aluminum cap with a tri-layer PTFE/silicone/PTFE septa and an 11 mm size, suitable for various laboratory instruments.
  • Cap color is blue.
  • Cap material is aluminum.
  • Cap size is 11 mm.
  • Septa layering is tri-layer.
  • Septa material is PTFE/silicone/PTFE.
  • Compatible with a wide range of instruments.
  • Suitable for GC and LC techniques.
  • Allows for repeat injections.
  • Designed for crimp vial types.
Catalog No. NC3981653 Supplier Agilent Technologies Supplier No. 51904074
This crimp cap is designed for GC and LC applications. It features a blue aluminum cap with a tri-layer PTFE/silicone/PTFE septa and an 11 mm size, suitable for various laboratory instruments.
  • Cap color is blue.
  • Cap material is aluminum.
  • Cap size is 11 mm.
  • Septa layering is tri-layer.
  • Septa material is PTFE/silicone/PTFE.
  • Compatible with a wide range of instruments.
  • Suitable for GC and LC techniques.
  • Allows for repeat injections.
  • Designed for crimp vial types.