Learn More
Agilent Technologies Agilent DNA 1000 Kit, Sufficient for 300 Samples
Catalog No. NC1758992
Encompass
For the separation, sizing and quantification of dsDNA fragments from 25 to 1000 bp. The kit includes 25 microfluidic chips, reagent and consumables, sufficient for 300 samples.
Catalog No. NC1758992 Supplier Agilent Technologies Supplier No. 50671504
For the separation, sizing and quantification of dsDNA fragments from 25 to 1000 bp. The kit includes 25 microfluidic chips, reagent and consumables, sufficient for 300 samples.