Learn More
Agilent Technologies AGILENT DNA 12000 KIT
Catalog No. NC1933217
Encompass
Agilent DNA 12000 Kit. For the separation, sizing and quantification of dsDNA fragments from 100 to 12000 bp. The kit includes 25 microfluidic chips, reagents and consumables, sufficient for 300 samples
Catalog No. NC1933217 Supplier Agilent Technologies Supplier No. 50671508
Agilent DNA 12000 Kit. For the separation, sizing and quantification of dsDNA fragments from 100 to 12000 bp. The kit includes 25 microfluidic chips, reagents and consumables, sufficient for 300 samples