Learn More
Agilent Technologies Agilent Ferrule, Graphite, 0.4 mm ID, 0.05 to 0.25 mm
Catalog No. NC3981656
Encompass
These graphite ferrules are designed for Gas Chromatography (GC) applications. They feature a 0.4 mm inner diameter and are compatible with 0.05 to 0.25 mm columns. They can be used in GC oven temperatures up to 450°C.
  • 0.4 mm inner diameter graphite ferrule
  • Compatible with 0.05 to 0.25 mm columns
  • For GC oven temperatures up to 450°C
  • Recommended for use with most selective GC detectors
  • Capillary ferrule type
  • Suitable for Gas Chromatography (GC) technique
Catalog No. NC3981656 Supplier Agilent Technologies Supplier No. 5002114
These graphite ferrules are designed for Gas Chromatography (GC) applications. They feature a 0.4 mm inner diameter and are compatible with 0.05 to 0.25 mm columns. They can be used in GC oven temperatures up to 450°C.
  • 0.4 mm inner diameter graphite ferrule
  • Compatible with 0.05 to 0.25 mm columns
  • For GC oven temperatures up to 450°C
  • Recommended for use with most selective GC detectors
  • Capillary ferrule type
  • Suitable for Gas Chromatography (GC) technique