Learn More
Agilent Technologies Agilent Fittings, Stainless Steel, Intermediate Long Screw, 1.6 mm ID, 10/Pack
Catalog No. NC3981639
Encompass
These fittings are constructed from stainless steel and feature an intermediate long screw design, suitable for various liquid chromatography applications. They are supplied in packs of 10 and include all necessary components for assembly.
  • Stainless steel construction ensures durability and chemical resistance.
  • Intermediate long screw (SI) design provides secure connections.
  • Includes stainless steel fitting screws, 11 mm in length, for reliable installation.
  • Comes with front and back ferrules for a complete sealing system.
  • Features an inner diameter of 1.6 mm for optimal flow.
  • Supplied in convenient packs of 10.
Catalog No. NC3981639 Supplier Agilent Technologies Supplier No. 50050068
These fittings are constructed from stainless steel and feature an intermediate long screw design, suitable for various liquid chromatography applications. They are supplied in packs of 10 and include all necessary components for assembly.
  • Stainless steel construction ensures durability and chemical resistance.
  • Intermediate long screw (SI) design provides secure connections.
  • Includes stainless steel fitting screws, 11 mm in length, for reliable installation.
  • Comes with front and back ferrules for a complete sealing system.
  • Features an inner diameter of 1.6 mm for optimal flow.
  • Supplied in convenient packs of 10.