Learn More
Agilent Technologies Agilent Gas Clean Purifier Kit, 8890/8860 GC, Sensor, Combination Filter, 1/8 in.
Catalog No. NC3994237
Encompass
This purification system for 8890 and 8860 GC systems delivers high-purity gases to analytical instruments, enhancing instrument life, sensitivity, data accuracy, and stability. It includes a mounting bracket, connecting unit, sensor, and carrier gas filter.
  • Delivers high-purity gases.
  • Handles high flow rates (up to 20 L/min).
  • Features fast-stabilizing, absorbent filter material.
  • Durable, transparent polycarbonate housing.
  • Tool-free filter replacement.
  • Automatic gas flow activation.
  • Sensor provides LED filter status and GC diagnostic notifications.
  • Protects GC components from oxidation and hydrolization.
  • Removes organic compounds, oxygen, moisture, and hydrocarbons.
  • Enhances GC productivity and faster stabilization.
  • Ensures high data accuracy and long-term stability.
Catalog No. NC3994237 Supplier Agilent Technologies Supplier No. CP179880
This purification system for 8890 and 8860 GC systems delivers high-purity gases to analytical instruments, enhancing instrument life, sensitivity, data accuracy, and stability. It includes a mounting bracket, connecting unit, sensor, and carrier gas filter.
  • Delivers high-purity gases.
  • Handles high flow rates (up to 20 L/min).
  • Features fast-stabilizing, absorbent filter material.
  • Durable, transparent polycarbonate housing.
  • Tool-free filter replacement.
  • Automatic gas flow activation.
  • Sensor provides LED filter status and GC diagnostic notifications.
  • Protects GC components from oxidation and hydrolization.
  • Removes organic compounds, oxygen, moisture, and hydrocarbons.
  • Enhances GC productivity and faster stabilization.
  • Ensures high data accuracy and long-term stability.