Learn More
Perkin Elmer US LLC Agilent Graphite Tubes with Omega Platform, Pkg. 10
Catalog No. 502111591
Encompass_Preferred
These tubes are fit to perform on atomic absorption (AA) instruments for Agilent and is equivalent to Agilent/Varian part 63-100037-00. PerkinElmer supplies a wide range of graphite tubes for non-PerkinElmer instrumentation. Graphite tubes are made with the highest quality material guaranteed to work for your AA instrument.
Catalog No. 50-211-1591 Supplier Perkin Elmer US LLC Supplier No. N9304628
Add to Cart
Add to Cart
Request a Quote
These tubes are fit to perform on atomic absorption (AA) instruments for Agilent and is equivalent to Agilent/Varian part 63-100037-00. PerkinElmer supplies a wide range of graphite tubes for non-PerkinElmer instrumentation. Graphite tubes are made with the highest quality material guaranteed to work for your AA instrument.