Learn More
Agilent Technologies InfinityLab Poroshell 120 EC-C18 UHPLC Guard Column, 2.7 μm, 2.1 mm x 5 mm, 3/Pk.
Catalog No. NC1394161
Encompass
These guard columns utilize superficially porous particle technology, featuring a solid silica core and a porous outer layer designed to enhance chromatographic efficiency and support rapid, high-resolution separations. The guard column helps protect analytical columns by trapping contaminants, thereby maximizing the lifetime of the analytical column. It is configured for reversed-phase applications.
  • Superficially porous particle architecture for improved performance
  • Designed to enable fast, high-resolution separations
  • Helps maximize the lifetime of analytical columns
  • Reversed-phase mode suitable for a wide range of applications
  • 2.7 μm particle size with 120 Å pore size
  • Includes three units per package
Catalog No. NC1394161 Supplier Agilent Technologies Supplier No. 821725911
These guard columns utilize superficially porous particle technology, featuring a solid silica core and a porous outer layer designed to enhance chromatographic efficiency and support rapid, high-resolution separations. The guard column helps protect analytical columns by trapping contaminants, thereby maximizing the lifetime of the analytical column. It is configured for reversed-phase applications.
  • Superficially porous particle architecture for improved performance
  • Designed to enable fast, high-resolution separations
  • Helps maximize the lifetime of analytical columns
  • Reversed-phase mode suitable for a wide range of applications
  • 2.7 μm particle size with 120 Å pore size
  • Includes three units per package