Learn More
Optimize Technologies Deuterium Lamp, Agilent/HP 1100 VWD G1314A
SDP
Catalog No. 502006622
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number G1314-60100.
Catalog No. 50-200-6622 Supplier Optimize Technologies Supplier No. 273202715
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number G1314-60100.