Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neta Scientific AGILENT PLRP-S 4000A, 4.6 X 15
SDP

Supplier:  Neta Scientific PL15123803

Encompass_Preferred

This item is manufactured by Agilent

Agilent PLRP-S 4000A, 4.6 x 150 mm, 8 um BC

Catalog No. NC2156994


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.