Learn More
Agilent Technologies RNA 6000 Pico Kit
Catalog No. NC1711873
Encompass
Agilent RNA 6000 Pico Kit; For the analysis of RNA samples of limited abundance down to 50 pg/µl of total RNA or 250 pg/µl of mRNA. The kit includes 25 microfluidic chips, reagents and consumables, sufficient for 275 samples.
Catalog No. NC1711873 Supplier Agilent Technologies Supplier No. 50671513
Agilent RNA 6000 Pico Kit; For the analysis of RNA samples of limited abundance down to 50 pg/µl of total RNA or 250 pg/µl of mRNA. The kit includes 25 microfluidic chips, reagents and consumables, sufficient for 275 samples.