Learn More
Agilent Technologies Agilent Small RNA kit
Supplier:  Agilent Technologies 50671548
Encompass
Agilent Small RNA kit. For the analysis and quantitation of small RNA samples from 6 to 150 nt in size and 50 to 2000 pg/L in concentration. The kit includes 25 microfluidic chips, reagents and consumables, sufficient for 275 samples.
Catalog No. NC1799518