Learn More
Agilent Technologies Agilent Technologies ALS Syringe, 10 μL, Fixed Needle, Tapered
Supplier:  Agilent Technologies 51813360
Encompass
This autosampler syringe is a Gold Standard design with a 10 μL volume and a tapered, fixed needle. It is primarily recommended for 1–8 μL injections in Gas Chromatography (GC) applications.
  • Tapered needle offers the strength of a 23-gauge in the upper portion and the versatility of a 26s-gauge in the lower portion.
  • Microliter syringe for precise injections.
  • Recommended for 1–8 μL injections.
  • Equipped with a stainless steel standard plunger.
  • Designed for use with split/splitless or on-column injections with 0.53 mm ID columns.
  • Provides durability due to the 23-gauge upper portion of the tapered needle.
  • Offers versatility for split/splitless and on-column injections with its 26s-gauge lower portion.
  • Suitable for autosamplers.
Catalog No. NC3981662