Learn More
Agilent Technologies Agilent Technologies Crimp Cap, 11 mm, Red, PTFE/Silicone/PTFE
Catalog No. NC3981654
Encompass
This crimp cap features a red aluminum cap and a tri-layer PTFE/silicone/PTFE septa, designed for secure sealing of 11 mm crimp vials. It is suitable for use in various analytical techniques including Gas Chromatography (GC) and Liquid Chromatography (LC).
  • Red cap color.
  • Aluminum cap material.
  • 11 mm cap size.
  • Includes septa.
  • Tri-layer septa.
  • PTFE/silicone/PTFE septa material.
  • Septa are not pre-slit.
  • Compatible with various autosamplers.
  • Suitable for repeat injections.
  • Designed for GC and LC techniques.
  • For crimp vials.
Catalog No. NC3981654 Supplier Agilent Technologies Supplier No. 51904075
This crimp cap features a red aluminum cap and a tri-layer PTFE/silicone/PTFE septa, designed for secure sealing of 11 mm crimp vials. It is suitable for use in various analytical techniques including Gas Chromatography (GC) and Liquid Chromatography (LC).
  • Red cap color.
  • Aluminum cap material.
  • 11 mm cap size.
  • Includes septa.
  • Tri-layer septa.
  • PTFE/silicone/PTFE septa material.
  • Septa are not pre-slit.
  • Compatible with various autosamplers.
  • Suitable for repeat injections.
  • Designed for GC and LC techniques.
  • For crimp vials.