Learn More
Agilent Technologies Agilent Technologies Inlet Septa, Non-stick, BTO, 11 mm, 400/pk
Catalog No. NC3981660
Encompass
These inlet septa are bleed and temperature optimized (BTO) and non-stick. They are designed for use with Split/Splitless (S/SL), Programmable Temperature Vaporizer (PTV), and Multi-Mode Injection (MMI) inlets. Septa come pre-conditioned and ready to use.
  • Non-stick
  • Bleed and temperature optimized (BTO)
  • Suitable for injection port temperatures up to 400 °C
  • Pre-conditioned, ready to use
Catalog No. NC3981660 Supplier Agilent Technologies Supplier No. 51903157
These inlet septa are bleed and temperature optimized (BTO) and non-stick. They are designed for use with Split/Splitless (S/SL), Programmable Temperature Vaporizer (PTV), and Multi-Mode Injection (MMI) inlets. Septa come pre-conditioned and ready to use.
  • Non-stick
  • Bleed and temperature optimized (BTO)
  • Suitable for injection port temperatures up to 400 °C
  • Pre-conditioned, ready to use